3-Methoxy-4-(2-phenylethoxy)benzaldehyde

Supplier: Matrix Scientific

030789-500MG
101830-832EA 387.69 USD
101830-832
3-Methoxy-4-(2-phenylethoxy)benzaldehyde
3-Methoxy-4-(2-phenylethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₃
MDL Number: MFCD03270635
CAS Number: 149428-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR