3-(2-Phenylethoxy)benzaldehyde

Supplier: Matrix Scientific

030787-500MG
101830-828EA 336.92 USD
101830-828
3-(2-Phenylethoxy)benzaldehyde
3-(2-Phenylethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MDL Number: MFCD09704955
CAS Number: 255832-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR