2-(2-Phenylethoxy)benzaldehyde

Supplier: Matrix Scientific

030786-500MG
101830-826EA 230.77 USD
101830-826
2-(2-Phenylethoxy)benzaldehyde
2-(2-Phenylethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₂
MDL Number: MFCD03820021
CAS Number: 93898-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR