4-[(2-Bromobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

030779-500MG
101830-812EA 387.69 USD
101830-812
4-[(2-Bromobenzyl)oxy]benzaldehyde
4-[(2-Bromobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁BrO₂
MDL Number: MFCD03765585
CAS Number: 632300-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR