4-(Allyloxy)-3-bromo-5-methoxybenzaldehyde

Supplier: Matrix Scientific

030777-500MG
101830-808EA 387.69 USD
101830-808
4-(Allyloxy)-3-bromo-5-methoxybenzaldehyde
4-(Allyloxy)-3-bromo-5-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₁BrO₃
MDL Number: MFCD02256557
CAS Number: 361465-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR