4-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde
Supplier: Matrix Scientific
030776-500MG
101830-806EA
387.69
USD
101830-806
4-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde
4-(Benzyloxy)-3-bromo-5-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₅H₁₃BrO₃ Melting Pt: 45…46 °C |
MDL Number:
MFCD01119124 CAS Number: 2556-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...