2-(Allyloxy)benzaldehyde

Supplier: Matrix Scientific

030774-500MG
101830-802EA 11.5 USD
101830-802
2-(Allyloxy)benzaldehyde
2-(Allyloxy)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 130 °C (10 Torr)
Density: 1.0938
MDL Number: MFCD00014130
CAS Number: 28752-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR