2-(2-Bromo-4,5-dimethoxyphenyl)-1,3-thiazolidine-4-carboxylic acid

Supplier: Matrix Scientific

030766-500MG
101830-786EA 387.69 USD
101830-786
2-(2-Bromo-4,5-dimethoxyphenyl)-1,3-thiazolidine-4-carboxylic acid
2-(2-Bromo-4,5-dimethoxyphenyl)-1,3-thiazolidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄BrNO₄S
MDL Number: MFCD09971996
CAS Number: 1101223-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR