(4-Amino-1,2,5-oxadiazol-3-yl)(phenyl)methanone

Supplier: Matrix Scientific

030764-500MG
101830-782EA 538.46 USD
101830-782
(4-Amino-1,2,5-oxadiazol-3-yl)(phenyl)methanone
(4-Amino-1,2,5-oxadiazol-3-yl)(phenyl)methanone

Hazard: Irritant

Formula: C₉H₇N₃O₂
MDL Number: MFCD00462566
CAS Number: 38924-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR