1-[1-(4-Amino-1,2,5-oxadiazol-3-yl)-1H-1,2,3-triazol-5-yl]ethanone

Supplier: Matrix Scientific

030763-500MG
101830-780EA 387.69 USD
101830-780
1-[1-(4-Amino-1,2,5-oxadiazol-3-yl)-1H-1,2,3-triazol-5-yl]ethanone
1-[1-(4-Amino-1,2,5-oxadiazol-3-yl)-1H-1,2,3-triazol-5-yl]ethanone

Hazard: Irritant

Formula: C₆H₆N₆O₂
MDL Number: MFCD09971994
CAS Number: 898745-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR