Ethyl-5-(chloromethyl)-1-[4-(1H-pyrrol-1-yl)-1,2,5-oxadiazol-3-yl]-1H-1,2,3-triazole-4-carboxylate

Supplier: Matrix Scientific

030762-500MG
101830-778EA 387.69 USD
101830-778
Ethyl-5-(chloromethyl)-1-[4-(1H-pyrrol-1-yl)-1,2,5-oxadiazol-3-yl]-1H-1,2,3-triazole-4-carboxylate
Ethyl-5-(chloromethyl)-1-[4-(1H-pyrrol-1-yl)-1,2,5-oxadiazol-3-yl]-1H-1,2,3-triazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁ClN₆O₃
MDL Number: MFCD03766116
CAS Number: 681851-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR