1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(methoxymethyl)1H-1,2,3-triazole-4-carboxylic acid

Supplier: Matrix Scientific

030759-500MG
101830-772EA 387.69 USD
101830-772
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(methoxymethyl)1H-1,2,3-triazole-4-carboxylic acid
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(methoxymethyl)1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₈N₆O₄
MDL Number: MFCD01414686
CAS Number: 312496-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR