1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(piperidin-1-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid

Supplier: Matrix Scientific

030758-500MG
101830-770EA 387.69 USD
101830-770
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(piperidin-1-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(piperidin-1-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₅N₇O₃
MDL Number: MFCD02130401
CAS Number: 352446-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR