1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(morpholin-4-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid

Supplier: Matrix Scientific

030757-500MG
101830-768EA 387.69 USD
101830-768
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(morpholin-4-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-(morpholin-4-ylmethyl)-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₃N₇O₄
MDL Number: MFCD01414692
CAS Number: 352446-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR