2-[(9-Phenyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]propanoic acid

Supplier: Matrix Scientific

030735-500MG
101830-728EA 387.69 USD
101830-728
2-[(9-Phenyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]propanoic acid
2-[(9-Phenyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]propanoic acid

Hazard: Irritant

Formula: C₁₇H₁₄N₄O₂S
MDL Number: MFCD06755489
CAS Number: 891449-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR