2-[(9-Benzyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]acetamide

Supplier: Matrix Scientific

030734-500MG
101830-726EA 387.69 USD
101830-726
2-[(9-Benzyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]acetamide
2-[(9-Benzyl-9H-[1,2,4]triazolo[4,3-a]benzimidazol-3-yl)thio]acetamide

Hazard: Irritant

Formula: C₁₇H₁₅N₅OS
MDL Number: MFCD04206107
CAS Number: 606120-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR