5-(2-Methyl-3-furyl)-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030355-500MG
101829-972EA 193.85 USD
101829-972
5-(2-Methyl-3-furyl)-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol
5-(2-Methyl-3-furyl)-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₂S
MDL Number: MFCD03423472
CAS Number: 725222-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR