4-Propyl-5-thien-2-yl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030353-500MG
101829-968EA 193.85 USD
101829-968
4-Propyl-5-thien-2-yl-4H-1,2,4-triazole-3-thiol
4-Propyl-5-thien-2-yl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₁N₃S₂
MDL Number: MFCD03423470
CAS Number: 451501-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR