4-Ethyl-5-thien-2-yl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030351-500MG
101829-964EA 193.85 USD
101829-964
4-Ethyl-5-thien-2-yl-4H-1,2,4-triazole-3-thiol
4-Ethyl-5-thien-2-yl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₉N₃S₂
MDL Number: MFCD03161622
CAS Number: 337473-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR