5-(5-Bromo-2-furyl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030345-500MG
101829-952EA 193.85 USD
101829-952
5-(5-Bromo-2-furyl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(5-Bromo-2-furyl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₇H₆BrN₃OS
MDL Number: MFCD03423465
CAS Number: 860790-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR