5-(2-Furyl)-4-isopropyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030344-500MG
101829-950EA 193.85 USD
101829-950
5-(2-Furyl)-4-isopropyl-4H-1,2,4-triazole-3-thiol
5-(2-Furyl)-4-isopropyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₁N₃OS
MDL Number: MFCD03423463
CAS Number: 588682-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR