5-(2-Furyl)-4-propyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030343-500MG
101829-948EA 193.85 USD
101829-948
5-(2-Furyl)-4-propyl-4H-1,2,4-triazole-3-thiol
5-(2-Furyl)-4-propyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₁N₃OS
MDL Number: MFCD03423462
CAS Number: 606110-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR