5-(2-Furyl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030342-500MG
101829-946EA 193.85 USD
101829-946
5-(2-Furyl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(2-Furyl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₇H₇N₃OS
MDL Number: MFCD00052247
CAS Number: 27106-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR