5-(1-Benzothien-3-yl)-4-propyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030339-500MG
101829-940EA 193.85 USD
101829-940
5-(1-Benzothien-3-yl)-4-propyl-4H-1,2,4-triazole-3-thiol
5-(1-Benzothien-3-yl)-4-propyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₁₃N₃S₂
MDL Number: MFCD03423459
CAS Number: 777879-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR