4-Allyl-5-(1-benzothien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030338-500MG
101829-938EA 193.85 USD
101829-938
4-Allyl-5-(1-benzothien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Allyl-5-(1-benzothien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₁₁N₃S₂
MDL Number: MFCD03423458
CAS Number: 522597-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR