5-(1-Benzothien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030336-500MG
101829-934EA 193.85 USD
101829-934
5-(1-Benzothien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(1-Benzothien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₁H₉N₃S₂
MDL Number: MFCD03423457
CAS Number: 588687-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR