4-Ethyl-5-(4,5,6,7-tetrahydro-1-benzothien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030327-500MG
101829-916EA 193.85 USD
101829-916
4-Ethyl-5-(4,5,6,7-tetrahydro-1-benzothien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Ethyl-5-(4,5,6,7-tetrahydro-1-benzothien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₂H₁₅N₃S₂
MDL Number: MFCD03075344
CAS Number: 438230-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR