4-Allyl-5-(4,5-dimethylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030318-500MG
101829-898EA 193.85 USD
101829-898
4-Allyl-5-(4,5-dimethylthien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Allyl-5-(4,5-dimethylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₁H₁₃N₃S₂
MDL Number: MFCD03423433
CAS Number: 522597-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR