5-(4,5-Dimethylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030316-500MG
101829-894EA 193.85 USD
101829-894
5-(4,5-Dimethylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(4,5-Dimethylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₁N₃S₂
MDL Number: MFCD03423432
CAS Number: 588685-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR