4-Allyl-5-(5-isopropylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030313-500MG
101829-888EA 193.85 USD
101829-888
4-Allyl-5-(5-isopropylthien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Allyl-5-(5-isopropylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₂H₁₅N₃S₂
MDL Number: MFCD03423429
CAS Number: 861226-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR