5-(5-Isopropylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030311-500MG
101829-884EA 193.85 USD
101829-884
5-(5-Isopropylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol
5-(5-Isopropylthien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₀H₁₃N₃S₂
MDL Number: MFCD03423428
CAS Number: 588687-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR