4-Methyl-5-(5-propylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030306-500MG
101829-874EA 193.85 USD
101829-874
4-Methyl-5-(5-propylthien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Methyl-5-(5-propylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₀H₁₃N₃S₂
MDL Number: MFCD03423424
CAS Number: 588687-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR