5-(5-Ethylthien-3-yl)-4-isopropyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030305-500MG
101829-872EA 193.85 USD
101829-872
5-(5-Ethylthien-3-yl)-4-isopropyl-4H-1,2,4-triazole-3-thiol
5-(5-Ethylthien-3-yl)-4-isopropyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₁H₁₅N₃S₂
MDL Number: MFCD03423422
CAS Number: 869952-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR