4-Allyl-5-(5-ethylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030303-500MG
101829-868EA 193.85 USD
101829-868
4-Allyl-5-(5-ethylthien-3-yl)-4H-1,2,4-triazole-3-thiol
4-Allyl-5-(5-ethylthien-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₁H₁₃N₃S₂
MDL Number: MFCD03423420
CAS Number: 522597-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR