2-Methyl-2-(4-nitrophenoxy)propanoic acid

Supplier: Matrix Scientific

028269-500MG
101825-814EA 146.15 USD
101825-814
2-Methyl-2-(4-nitrophenoxy)propanoic acid
2-Methyl-2-(4-nitrophenoxy)propanoic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₅
MDL Number: MFCD03422282
CAS Number: 17431-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR