2-Methyl-2-(2-naphthyloxy)propanoic acid

028267-500MG
101825-810EA 228.85 USD
101825-810
2-Methyl-2-(2-naphthyloxy)propanoic acid
2-Methyl-2-(2-naphthyloxy)propanoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₃
MDL Number: MFCD01570469
CAS Number: 7756-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR