2-(4-Bromophenoxy)-2-methylpropanoic acid
Supplier: Matrix Scientific
028261-500MG
101825-798EA
158.46
USD
101825-798
2-(4-Bromophenoxy)-2-methylpropanoic acid
2-(4-Bromophenoxy)-2-methylpropanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₁BrO₃ |
MDL Number:
MFCD01995843 CAS Number: 7472-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...