2-(2-Chloro-5-methylphenoxy)-2-methylpropanoic acid

Supplier: Matrix Scientific

028252-500MG
101825-780EA 146.15 USD
101825-780
2-(2-Chloro-5-methylphenoxy)-2-methylpropanoic acid
2-(2-Chloro-5-methylphenoxy)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃ClO₃
MDL Number: MFCD03422264
CAS Number: 588692-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR