2-(1,1'-Biphenyl-4-yloxy)butanoic acid

Supplier: Matrix Scientific

028250-500MG
101825-776EA 146.15 USD
101825-776
2-(1,1'-Biphenyl-4-yloxy)butanoic acid
2-(1,1'-Biphenyl-4-yloxy)butanoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₃
MDL Number: MFCD03422262
CAS Number: 101169-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR