2-(4-Chloro-3-methylphenoxy)butanoic acid

Supplier: Matrix Scientific

028248-500MG
101825-772EA 146.15 USD
101825-772
2-(4-Chloro-3-methylphenoxy)butanoic acid
2-(4-Chloro-3-methylphenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃ClO₃
MDL Number: MFCD03422260
CAS Number: 869947-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR