2-(2-Naphthyloxy)butanoic acid

Supplier: Matrix Scientific

028240-500MG
101825-756EA 146.15 USD
101825-756
2-(2-Naphthyloxy)butanoic acid
2-(2-Naphthyloxy)butanoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₃
MDL Number: MFCD00757431
CAS Number: 161904-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR