2-(2,5-Dichlorophenoxy)butanoic acid

Supplier: Matrix Scientific

028234-500MG
101825-744EA 146.15 USD
101825-744
2-(2,5-Dichlorophenoxy)butanoic acid
2-(2,5-Dichlorophenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀Cl₂O₃
MDL Number: MFCD03422247
CAS Number: 855152-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR