2-(4-Chlorophenoxy)butanoic acid

Supplier: Matrix Scientific

028232-500MG
101825-740EA 307.69 USD
101825-740
2-(4-Chlorophenoxy)butanoic acid
2-(4-Chlorophenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₁ClO₃
MDL Number: MFCD02295734
CAS Number: 34385-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR