2-(3-Methyl-4-nitrophenoxy)butanoic acid

Supplier: Matrix Scientific

028230-500MG
101825-736EA 146.15 USD
101825-736
2-(3-Methyl-4-nitrophenoxy)butanoic acid
2-(3-Methyl-4-nitrophenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₅
MDL Number: MFCD03422239
CAS Number: 869948-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR