2-(4-Bromo-2-chlorophenoxy)butanoic acid

Supplier: Matrix Scientific

028226-500MG
101825-728EA 116.92 USD
101825-728
2-(4-Bromo-2-chlorophenoxy)butanoic acid
2-(4-Bromo-2-chlorophenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀BrClO₃
MDL Number: MFCD03422234
CAS Number: 861507-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR