2-(2-Isopropyl-5-methylphenoxy)propanoic acid

Supplier: Matrix Scientific

028222-500MG
101825-720EA 146.15 USD
101825-720
2-(2-Isopropyl-5-methylphenoxy)propanoic acid
2-(2-Isopropyl-5-methylphenoxy)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₈O₃
MDL Number: MFCD03422230
CAS Number: 159302-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR