2-[4-(1-Methyl-1-phenylethyl)phenoxy]propanoic acid

Supplier: Matrix Scientific

028220-500MG
101825-716EA 146.15 USD
101825-716
2-[4-(1-Methyl-1-phenylethyl)phenoxy]propanoic acid
2-[4-(1-Methyl-1-phenylethyl)phenoxy]propanoic acid

Hazard: Irritant

Formula: C₁₈H₂₀O₃
MDL Number: MFCD03422228
CAS Number: 70757-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR