2-(4-Bromophenoxy)propanoic acid
Supplier: Matrix Scientific
028214-500MG
101825-704EA
146.15
USD
101825-704
2-(4-Bromophenoxy)propanoic acid
2-(4-Bromophenoxy)propanoic acid
Hazard: Irritant
Formula:
C₉H₉BrO₃ |
MDL Number:
MFCD02295723 CAS Number: 32019-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...