2-(4-Bromo-2-chlorophenoxy)propanoic acid

Supplier: Matrix Scientific

028208-500MG
101825-692EA 146.15 USD
101825-692
2-(4-Bromo-2-chlorophenoxy)propanoic acid
2-(4-Bromo-2-chlorophenoxy)propanoic acid

Hazard: Irritant

Formula: C₉H₈BrClO₃
MDL Number: MFCD03422212
CAS Number: 588681-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR