6-Nitro-2-oxo-2H-chromene-3-carboxylic acid

Supplier: Matrix Scientific

028195-500MG
101825-666EA 336.92 USD
101825-666
6-Nitro-2-oxo-2H-chromene-3-carboxylic acid
6-Nitro-2-oxo-2H-chromene-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅NO₆
MDL Number: MFCD00052754
CAS Number: 10242-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR